missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RC Antibody (309822), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB22691Y
This item is not returnable.
View return policy
Description
IL-17RC Monoclonal antibody specifically detects IL-17RC in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
IL-17RC | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
84818 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, CyTOF | |
309822 | |
Flow Cytometry, CyTOF-ready | |
FLJ95963, FLJ96005, IL-17 receptor C, IL17F receptor, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14 | |
Mouse myeloma cell line NS0-derived recombinant human IL-17 RC, Leu21-Ala454, Accession # NP_116121 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction