missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RD/SEF Antibody (001), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89540F
This item is not returnable.
View return policy
Description
IL-17RD/SEF Monoclonal antibody specifically detects IL-17RD/SEF in Human samples. It is validated for ELISA, ELISA
Specifications
| IL-17RD/SEF | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 54756 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 001 | |
| ELISA, Sandwich ELISA | |
| FLJ35755, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, Sef homolog, SEFhSef | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-17RD/SEF (NP_060033.3; Met1-Arg299). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction