missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 R beta Antibody (Singapore ASTR patent anti-IL-2R beta/ IL-2R gamma) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28336JF525
This item is not returnable.
View return policy
Description
IL-2 R beta Monoclonal antibody specifically detects IL-2 R beta in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IL-2 R beta | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
| IL-2Rb/ CD122 | |
| 0.1 mL | |
| Immunology | |
| 3560 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Singapore ASTR patent anti-IL-2R beta / IL-2R gamma | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction