Learn More
Novus Biologicals™ IL-21 Antibody (Lilly patent anti-IL-21) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28012-100ug
 View more versions of this product
Product Code. 30048714
  • 338.00€ / 100µg
Description
IL-21 Monoclonal antibody specifically detects IL-21 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-21
Monoclonal
Unconjugated
Q9HBE4
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Lilly patent anti-IL-21
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform
IL-21
100 μg
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction
59067