Learn More
IL-3 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 500-P23BT-1MG
Description
AA Sequence of recombinant protein: APMTQTTSLK TSWVNCSNMI DEIITHLKQP PLPLLDFNNL NGEDQDILME NNLRRPNLEA FNRAVKSLQN ASAIESILKN LLPCLPLATA APTRHPIHIK DGDWNEFRRK LTFYLKTLEN AQAQQTTLSL AIF. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-3. Anti-Human IL-3-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hIL-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-3 (500-P23) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hIL-3. Western Blot: To detect hIL-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIL-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P23BT-1MG will be provided as 2 x 500 μg
Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
Specifications
| IL-3 | |
| Polyclonal | |
| Biotin | |
| IL3 | |
| BP/BPA; BPA; Colony stimulating factor multiple; colony-stimulating factor, multiple; Csfmu; Eo-CSF; HCGF; hematopoietic growth factor; Il3; Il-3; ILN; Interleukin; interleukin 3; Interleukin 3 (colony stimulating factor; Interleukin3; interleukin-3; Mast cell growth factor; mast-cell growth factor; MCGF; MGC79398; MGC79399; MGF; Multi CSF; MULTI-CSF; Multilineage colony stimulating factor; multilineage-colony-stimulating factor; multiple); multipotential colony-stimulating factor; P cell stimulating factor; P-cell stimulating factor; P-cell-stimulating factor; PCSA; PSF | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3562 | |
| -20°C |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P08700 | |
| IL3 | |
| E.coli-derived Recombinant Human IL-3 | |
| 2 x 500μg | |
| Primary | |
| Human | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.