missing translation for 'onlineSavingsMsg'
Learn More

IL-4 Rat anti-Mouse, APC, Clone: 11B11, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 554436

 View more versions of this product

Product Code. 15867698

  • ONLINE EXCLUSIVE PRICE
    305.00€
    303.88€ / Each
    Save 1.12€
    0.37% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 11B11 antibody reacts with mouse interleukin-4 (IL-4). The immunogen used to generate the 11B11 hybridoma was partially purified mouse IL-4 from PMA-stimulated EL-4 supernatant. This is a neutralizing antibody.

Host Species: Rat
Clone: 11B11
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Partially Purified Mouse IL-4

Intracellular Staining

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-4
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Partially Purified Mouse IL-4
0.1 mg
Immunology
Mouse
IgG1
Flow Cytometry
11B11
APC
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.