Learn More
IL-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 39 publications
Brand:  Novus Biologicals NB600-1131
 View more versions of this product
Product Code. 18790664
  • 532.00€ / 0.20mL
Description
IL-6 Polyclonal specifically detects IL-6 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, ELISA, Immunohistochemistry (Frozen), KnockDown
77 mg/mL
B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6