missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6R alpha Antibody (033), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90512AF350
This item is not returnable.
View return policy
Description
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| IL-6R alpha | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3570 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunofluorescence | |
| 33 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CD126, gp80, IL6Q, IL6R, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-6R alpha (NP_034689.2; Ile44-Glu357). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu