Learn More
IL-7R alpha/CD127 Antibody (717519) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8484F
Product Code. 18624085
  • 577.00€ / 0.10mL
Description
IL-7R alpha/CD127 Monoclonal antibody specifically detects IL-7R alpha/CD127 in Rat samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-7R alpha/CD127
Monoclonal
FITC
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Rat
Purified
Flow Cytometry, CyTOF
717519
Flow Cytometry, CyTOF-ready
CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Mouse myeloma cell line NS0-derived recombinant rat IL-7 R alpha/CD127, Glu21-Asp239, Accession # NP_001099888
0.1 mL
Apoptosis, Cytokine Research, Immunology, Innate Immunity
3575
Store at 4°C in the dark.
IgG2a