Learn More
Invitrogen™ IL-9 Recombinant Rabbit Monoclonal Antibody (016)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560527
Product Code. 30536217
  • 429.00€ / 100µL
Description
IL-9 Recombinant Monoclonal Antibody for ELISA
Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-9
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P15247
Il9
Recombinant Mouse IL-9 protein (Met1-Pro144).
100 μL
Primary
Mouse
Antibody
IgG
ELISA
016
Unconjugated
Il9
cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40
Rabbit
Protein A
RUO
16198
-20°C, Avoid Freeze/Thaw Cycles