Learn More
Invitrogen™ IL12A/IL12B Recombinant Rabbit Monoclonal Antibody (068)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA530719
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human IL12A & IL12B Heterodimer.
Specifications
| IL12A/IL12B | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P29459, P29460 | |
| IL12A, IL12B | |
| Recombinant Human Human IL12A & IL12B Heterodimer protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 068 | |
| Unconjugated | |
| IL12A | |
| CLMF; CLMF p35; CLMF p40; CLMF2; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il12a; IL-12A; IL12B; Il-12b; IL-12p35; Il12p40; Il-12p40; IL35 subunit; IMD28; IMD29; Interleukin; interleukin 12, p35; interleukin 12, p40; interleukin 12a; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12B; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin-12 alpha chain; interleukin-12 beta chain; interleukin-12 subunit alpha; Interleukin-12 subunit beta; Ll12a; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; p35; p40 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3592, 3593 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.