missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL17D Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  PeproTech 500-P88-100UG

 View more versions of this product

Product Code. 17863413

  • 284.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: APRAGRRPAR PRGCADRPEE LLEQLYGRLA AGVLSAFHHT LQLGPREQAR NASCPAGGRP ADRRFRPPTN LRSVSPWAYR ISYDPARYPR YLPEAYCLCR GCLTGLFGEE DVRFRSAPVY MPTVVLRRTP ACAGGRSVYT EAYVTIPVGC TCVPEPEKDA DSINSSIDKQ GAKLLLGPND APAGP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-17D. Anti-Human IL-17D-specific antibody was purified by affinity chromatography employing an immobilized Human IL-17D matrix. Sandwich ELISA: To detect Human IL-17D by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IL-17D (500-P88Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-17D. Western Blot: To detect Human IL-17D by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human IL-17D is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. The treatment of endothelial cells with this cytokine has been shown to stimulate the production of other cytokines including IL6, IL8 and CSF2/ GM-CSF. The increased expression of IL8 induced by this cytokine was found to be NF-kappa B-dependent.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL17D
Polyclonal
Unconjugated
IL17D
AI462269; FLJ30846; IL17D; IL-17D; IL-22; IL-27; Il27a; IL-3R; Interleukin; interleukin 17D; interleukin 27A; interleukin-17D; Interleukin-27; UNQ3096/PRO21175
Rabbit
Antigen affinity chromatography
RUO
53342
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
Q8TAD2
IL17D
E.coli-derived Recombinant Human IL-17D.
100 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.