Learn More
IL17RD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531871
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7, rat heart. Predicted reactivity: Mouse (92%), Rat (90%), Chicken (81%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Feedback inhibitor of fibroblast growth factor mediated Ras-MAPK signaling and ERK activation. May inhibit FGF-induced FGFR1 tyrosine phosphorylation. Regulates the nuclear ERK signaling pathway by spatially blocking nuclear translocation of activated ERK without inhibiting cytoplasmic phosphorylation of ERK. Mediates JNK activation and may be involved in apoptosis. Might have a role in the early stages of fate specification of GnRH-secreting neurons.
Specifications
| IL17RD | |
| Polyclonal | |
| Unconjugated | |
| Il17rd | |
| 2810004A10Rik; AI428510; HH18; hSef; IL-17 receptor D; IL17RD; IL-17RD; IL17Rhom; IL17RLM; interleukin 17 receptor D; interleukin-17 receptor D; Interleukin-17 receptor-like protein; mSef; RGD1565237; SEF; sef homolog; Sef-S; UNQ6115/PRO20026 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 498576, 54756 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8NFM7 | |
| Il17rd | |
| Recombinant protein fragment corresponding to a region within amino acids 1 and 209 of Human IL17 Receptor D | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.