missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL18R1 Monoclonal Antibody (112614)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA560945

Product Code. 30535580

  • 589.00€ / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL18R1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q61098
Il18r1
Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha /IL-1 R5, Ser19-Gly326.
500 μg
Primary
Mouse
Antibody
IgG1
Flow Cytometry, Western Blot
112614
Unconjugated
Il18r1
CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha
Rat
Protein A/G
RUO
16182
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.