missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL1RAPL1 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149498
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the interleukin 1 receptor family and is similar to the interleukin 1 accessory proteins. This protein has an N-terminal signal peptide, three extracellular immunoglobulin Ig-like domains, a transmembrane domain, an intracellular Toll/IL-1R domain, and a long C-terminal tail which interacts with multiple signaling molecules. This gene is located at a region on chromosome X that is associated with a non-syndromic form of X-linked intellectual disability. Deletions and mutations in this gene were found in patients with intellectual disability. This gene is expressed at a high level in post-natal brain structures involved in the hippocampal memory system, which suggests a specialized role in the physiological processes underlying memory and learning abilities, and plays a role in synapse formation and stabilization.
Specifications
| IL1RAPL1 | |
| Polyclonal | |
| Unconjugated | |
| Il1rapl1 | |
| 6330532G10Rik; C030039N24; IL1R8; Il1rapl; Il1rapl1; IL-1-RAPL-1; IL-1RAPL-1; IL1RAPL-1; interleukin 1 receptor accessory protein like 1; interleukin 1 receptor accessory protein-like 1; interleukin 1 receptor-8; interleukin-1 receptor accessory protein-like 1; mental retardation, X-linked 10; MRX10; MRX21; MRX34; Oligophrenin-4; OPHN4; Three immunoglobulin domain-containing IL-1 receptor-related 2; TIGIRR-2; X-linked interleukin-1 receptor accessory protein-like 1 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 11141 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9NZN1 | |
| Il1rapl1 | |
| Mouse myeloma cell line NS0-derived recombinant human IL1RAPL1, Leu19-Thr357. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction