missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL36RN Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13113-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine was shown to specifically inhibit the activation of NF-kappaB induced by interleukin 1 family, member 6 (IL1F6). This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Two alternatively spliced transcript variants encoding the same protein have been reported.Specifications
| IL36RN | |
| Polyclonal | |
| Unconjugated | |
| IL36RN | |
| FIL1, FIL1 delta, FIL1(DELTA), FIL1D, IL 1 delta, IL 1 related protein 3, IL 1F5, IL 1HY1, IL 1L1, IL 1ra homolog 1, IL 1RP3, IL1F5, IL1HY1, IL1L1, IL1RP3, Interleukin 1 delta, Interleukin 1 family member 5, Interleukin 1 HY1, Interleukin 1 like protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26525 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UBH0 | |
| IL36RN | |
| IL36RN Fusion Protein Ag3780 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction