Learn More
PeproTech IL4R (soluble) Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
219.00€ - 1834.00€
Specifications
| Antigen | IL4R (soluble) |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17875223
|
PeproTech
500-P325-50UG |
50 μg |
219.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17865213
|
PeproTech
500-P325-100UG |
100 μg |
275.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17865823
|
PeproTech
500-P325-1MG |
1 mg |
1834.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: GNMKVLQEPT CVSDYMSIST CEWKMNGPTN CSTELRLLYQ LVFLLSEAHT CIPENNGGAG CVCHLLMDDV VSADNYTLDL WAGQQLLWKG SFKPSEHVKP RAPGNLTVHT NVSDTLLLTW SNPYPPDNYL YNHLTYAVNI WSENDPADFR IYNVTYLEPS LRIAASTLKS GISYRARVRA WAQCYNTTWS EWSPSTKWHN SYREPFEQH. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human sIL-4 Receptor alpha. Anti-Human sIL-4 Receptor alpha-specific antibody was purified by affinity chromatography employing an immobilized Human sIL-4 Receptor alpha matrix. Sandwich ELISA: To detect Human sIL-4 Receptor alpha by sandwich ELISA (using 100 μL/well), a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human sIL-4 Receptor alpha (500-P325BT) as a detection antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human sIL-4 Receptor alpha. Western Blot: To detect Human sIL-4 Receptor alpha by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human sIL-4 Receptor alpha is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
IL4 Receptor alpha (IL4RA) is the alpha chain of the interleukin 4 receptor which binds to both interleukin 4 and interleukin 13 to regulate IgE production, chemokine and mucus production at sites of allergic inflammmation. The IL4 response is involved in promoting Th2 differentiation. The secreted extracellular domain of IL4R alpha, called sIL4R alpha, can inhibit IL4-mediated cell proliferation and IL5 upregulation by T-cells. It has no signaling abilities. Allelic variations in the IL4RA gene have been associated with atopy, a condition that can manifest itself as allergic rhinitis, sinusitus, asthma, or eczema.Specifications
| IL4R (soluble) | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P24394 | |
| 3566 | |
| CHO cells-derived Recombinant Human IL-4 Receptor alpha. | |
| Antigen affinity chromatography | |
| IL4R |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| 582J2.1; CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; Il4r; IL4R nirs variant 1; IL-4R subunit alpha; IL4RA; IL-4RA; IL-4R-alpha; interleukin 4 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha | |
| IL4R | |
| Primary | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.