missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL6R Antibody (AP-MAB0848) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
170.00€
Specifications
| Antigen | IL6R |
|---|---|
| Clone | AP-MAB0848 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18572601
|
Novus Biologicals
NBP1-06670-0.1mg |
0.1 mg | |||||||
Description
IL6R Monoclonal specifically detects IL6R in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF-ready, Block/Neutralize.Specifications
| IL6R | |
| 1 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Cancer, Immunology, Innate Immunity | |
| PBS with No Preservative | |
| CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991 | |
| IL6R | |
| IgG2 | |
| Protein A or G purified |
| AP-MAB0848 | |
| Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| P22272 | |
| 3570 | |
| Mouse recombinant IL-6R alpha chain | |
| Primary | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title