missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL9 Mouse anti-Human, Clone: 1F3A11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66144-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness.Specifications
| IL9 | |
| Monoclonal | |
| 1.63 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15248 | |
| IL9 | |
| IL9 Fusion Protein Ag21435 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1F3A11 | |
| Unconjugated | |
| IL9 | |
| Cytokine P40, HP40, IL 9, IL9, interleukin 9, P40, T cell growth factor P40 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3578 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction