missing translation for 'onlineSavingsMsg'
Learn More

Imatinib Hydrochloride, TRC

Product Code. 30134222
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134222 1 g 1g
30133903 500 mg 500mg
30009235 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134222 Supplier Toronto Research Chemicals Supplier No. TRCI2679801G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 862366-25-4
Molecular Formula C29 H31 N7 O . Cl H
Molecular Weight (g/mol) 530.06
Synonym Benzamide, 4-[(4-methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]-, hydrochloride (1:1), Benzamide, 4-[(4-methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]-, monohydrochloride (9CI), 4-[[4-Methyl-1-piperazinyl]methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide hydrochloride, 4-(4-Methylpiperazin-1-ylmethyl)-N-[4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]phenyl]benzamide hydrochloride, Genfatinib hydrochloride, Imatinib hydrochloride
IUPAC Name 4-[(4-methylpiperazin-1-yl)methyl]-N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide;hydrochloride
SMILES O=C(C1=CC=C(CN2CCN(CC2)C)C=C1)NC3=CC=C(C(NC4=NC=CC(C5=CC=CN=C5)=N4)=C3)C.[xHCl]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C29H31N7O.ClH/c1-21-5-10-25(18-27(21)34-29-31-13-11-26(33-29)24-4-3-12-30-19-24)32-28(37)23-8-6-22(7-9-23)20-36-16-14-35(2)15-17-36;/h3-13,18-19H,14-17,20H2,1-2H3,(H,32,37)(H,31,33,34);1H
Formula Weight 529.2357
Chemical Name or Material Imatinib Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.