missing translation for 'onlineSavingsMsg'
Learn More

Imatinib N1,N4-Dioxide, Mikromol™

Product Code. 30049680
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049680 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049680 Supplier Mikromol Supplier No. MM3566.210025
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Imatinib N1,N4-Dioxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imatinib N1,N4-dioxide
CAS 571186-93-1
Analyte or Component Names Imatinib N1,N4-dioxide
Shipping Condition Ice pack (-20°C)
Molecular Formula C29 H31 N7 O3
Synonym 4-[(4-methyl-1,4-dioxido-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide, Imatinib (Piperidine)-N,N-dioxide, Imatinib N1,N4-Dioxide, Benzamide, 4-[(4-methyl-1,4-dioxido-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]-
InChI Formula InChI=1S/C29H31N7O3/c1-21-5-10-25(18-27(21)34-29-31-13-11-26(33-29)24-4-3-12-30-19-24)32-28(37)23-8-6-22(7-9-23)20-36(39)16-14-35(2,38)15-17-36/h3-13,18-19H,14-17,20H2,1-2H3,(H,32,37)(H,31,33,34)
SMILES Cc1ccc(NC(=O)c2ccc(C[N+]3([O-])CC[N+](C)([O-])CC3)cc2)cc1Nc4nccc(n4)c5cccnc5
IUPAC Name 4-[(4-methyl-1,4-dioxidopiperazine-1,4-diium-1-yl)methyl]-N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide
Molecular Weight (g/mol) 525.6
Quantity 25 mg
Formula Weight 525.2488 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.