missing translation for 'onlineSavingsMsg'
Learn More

Imatinib (Piperidine)-N,N-dioxide, TRC

Product Code. 17944125
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17944125 1 mg 1mg
17911935 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17944125 Supplier Toronto Research Chemicals Supplier No. TRCI2680201MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 571186-93-1
Molecular Formula C29 H31 N7 O3
Molecular Weight (g/mol) 525.6
Synonym 4-[(4-methyl-1,4-dioxido-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide, Imatinib (Piperidine)-N,N-dioxide, Imatinib N1,N4-Dioxide, Benzamide, 4-[(4-methyl-1,4-dioxido-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]-
IUPAC Name 4-[(4-methyl-1,4-dioxidopiperazine-1,4-diium-1-yl)methyl]-N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide
SMILES Cc1ccc(NC(=O)c2ccc(C[N+]3([O-])CC[N+](C)([O-])CC3)cc2)cc1Nc4nccc(n4)c5cccnc5

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C29H31N7O3/c1-21-5-10-25(18-27(21)34-29-31-13-11-26(33-29)24-4-3-12-30-19-24)32-28(37)23-8-6-22(7-9-23)20-36(39)16-14-35(2,38)15-17-36/h3-13,18-19H,14-17,20H2,1-2H3,(H,32,37)(H,31,33,34)
Formula Weight 525.2488
Chemical Name or Material Imatinib N1,N4-dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.