missing translation for 'onlineSavingsMsg'
Learn More

Imazalil, TRC

Product Code. 30137562
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137562 10 g 10g
18063200 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137562 Supplier Toronto Research Chemicals Supplier No. TRCI26850310G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35554-44-0
Molecular Formula C14 H14 Cl2 N2 O
Molecular Weight (g/mol) 297.18
Synonym Enilconazole, Imazalil, 1-[(2RS)-2-(2,4-Dichlorophenyl)-2-(prop-2-enyloxy)ethyl]-1H-imidazole, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-(2-propen-1-yloxy)ethyl]-, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-(2-propenyloxy)ethyl]- (9CI), 1-[2-(2,4-Dichlorophenyl)-2-(2-propen-1-yloxy)ethyl]-1H-imidazole, (R,S)-1-[2-(2,4-Dichlorophenyl)-2-(2-propenyloxy)ethyl]-1H-imidazole, (RS)-1-[2-(Allyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole, (±)-Imazalil, 1-[2-(2,4-Dichlorophenyl)-2-(2-propenyloxy)ethyl]-1H-imidazole, Amolden MP 100, Bromazil, CGA 41333, Clinafarm, Deccosil, Deccozil, Deccozil S 75, Enilconazole, FloPro IMZ, Florasan, Freshgard, Fungaflor, Fungazil, Imaverol, Magnate, NuZone, R 23979
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-prop-2-enoxyethyl]imidazole
SMILES Clc1ccc(C(Cn2ccnc2)OCC=C)c(Cl)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H14Cl2N2O/c1-2-7-19-14(9-18-6-5-17-10-18)12-4-3-11(15)8-13(12)16/h2-6,8,10,14H,1,7,9H2
Formula Weight 296.0483
Chemical Name or Material Imazalil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.