missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridin-6-ylmethanol, 97%, BLD Pharm™

Product Code. 31089976
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31089976 25 g
31081782 100 mg
31087932 5 g
31087174 250 mg
31084624 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31089976 Supplier BLD Pharm Supplier No. BD13439025g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 132213-07-1
Molecular Formula C8H8N2O
MDL Number MFCD07368521
Quantity 25 g
SMILES N1=C2[N](C=C1)C=C(C=C2)CO
Molecular Weight (g/mol) 148.16
Percent Purity 97%
Chemical Name or Material Imidazo[1,2-a]pyridin-6-ylmethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.