missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-6-carbonitrile, ≥97%, Thermo Scientific™

Product Code. 10137892
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10137892 1 g 1g
10567853 250 mg 250mg
10158694 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10137892 Supplier Thermo Scientific Maybridge Supplier No. CC33116DA
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 106850-34-4
Molecular Formula C8H5N3
Molecular Weight (g/mol) 143.149
MDL Number MFCD06659603
InChI Key LRJOKNYELSECDZ-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carbonitrile, 6-cyanoimidazo 1,2-a pyridine, 4-hydroimidazo 1,2-a pyridine-6-carbonitrile, 6-cyano-imidazo 1,2-a pyridine, acmc-1bu34, 6-imidazo 1,2-a pyridinecarbonitrile, imidazo 1,2-, a pyridine-6-carbonitrile, imidazo 1,2-alpha pyridine-6-carbonitrile, 6-cyanoimidazo 1,2-a pyridine-6-carbonitrile
PubChem CID 2764471
IUPAC Name imidazo[1,2-a]pyridine-6-carbonitrile
SMILES C1=CC2=NC=CN2C=C1C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 143.15
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridine-6-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.