missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Immepip dihydrobromide

Description
Immepip dihydrobromide is a potent histamine H3 receptor agonist. Also binds to H4 receptors (Ki values are 0.4 and 9 nM at human recombinant H3 and H4 receptors respectively). Equipotent to or slightly more active than (R)-α-methylhistamine at H3 receptors.
Specifications
Specifications
| Chemical Name or Material | 4-(1H-Imidazol-4-ylmethyl)piperidine dihydrobromide |
| CAS | 164391-47-3 |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in water |
| Molecular Formula | C9H15N3.2HBr |
| Formula Weight | Observed MW: 327.06 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?