Learn More
Merck Immobilon™-E Transfer Membrane
Product Code. p-7114685 Shop All Merck Products
Change view
Click to view available options
Quantity:
1 Roll
10 Sheets
4 Sheets
50 Sheets
Dimensions (LxW):
1.875 m x 26.5 cm
10 m x 8.5 cm
10 x 10 cm
7 x 8.4 cm
8 x 10 cm
9 x 12 cm
Unit Size:
1 roll
Each
Pack of 10
Pack of 4
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Dimensions (LxW) unitSize
15867541 1 Roll 1.875 m x 26.5 cm 1 roll
16379591 1 Roll 10 m x 8.5 cm Each
15897531 4 Sheets 7 x 8.4 cm Pack of 4
15807541 50 Sheets 7 x 8.4 cm Pack of 50
15817541 10 Sheets 8 x 10 cm Pack of 10
15827541 10 Sheets 9 x 12 cm Pack of 10
15837541 10 Sheets 10 x 10 cm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15867541 Supplier Merck Supplier No. IEVH00005
Add to basket
Add to basket
Immobilon™-E provides the a PVDF blotting membrane that wets in transfer buffer or water and eliminates the need for the alcohol pre-wet step.
Features:
  • 0.45 μm pore size is ideal for most Western blotting applications with chemiluminescence or chromogenic detection
  • Saves time and reduces waste by eliminating the alcohol pre-wet step
  • Higher protein retention delivers stronger signal than most nitrocellulose membranes
TRUSTED_SUSTAINABILITY
Specifications
Description Easily wetted with water
For Use With (Application) Western Blotting
Color White
Code IEVH
Surface Plain
Quantity 1 Roll
Format Roll
Material PVDF
Pore Size 0.45 μm
Dimensions (LxW) 1.875 m x 26.5 cm