Learn More
Honeywell™ Impact Pro Amplification Earmuff
Product Code. 15886825
Change view
Click to view available options
Black, NNR: 30, SNR: 33
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit
15886825 Black, NNR: 30, SNR: 33
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15886825 Fournisseur Honeywell™ Code fournisseur 1018953
  • ONLINE EXCLUSIVE PRICE
    259.35€
    186.69€ / 1 pairs
    Save 72.66€
    28% Off
Estimated Shipment: 23-11-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Designed for handgun and pistol shooters looking for a high noise attenuation rating.
  • Voice amplification for communication on the range
  • Automatic 4-hour shut-off increases battery life
  • Connects to MP3 players, smart phones, and scanner radios
  • External battery compartment
  • Rubberized pressure points will prevent gunstock scratching
  • One single power and volume control knob
  • Aggressive tactical styling
TRUSTED_SUSTAINABILITY
Spécification
Certifications/Compliance EU Directives: 89/686/EEC, EC Category PPE: Category II, Standards: EN 352-1:2002, EN 352-4:2001, EN 352-8:2008, ISO 4869-1:1990, ISO 4869-2:1994, ISO 4869-3:2007, Laboratory: PZT GmbH, An der Junkerei 48 F D-26389 Wilhelmshaven Germany, Notified Body Number: 1974
For Use With (Application) Government, law enforcement, military, noise cancellation, firearms
Noise Reduction Rating (NRR) NNR: 30 dB, SNR: 33 dB
Includes 2 AAA batteries and 3.5 audio input connection cord