missing translation for 'onlineSavingsMsg'
Learn More

Imperatorin, TRC

Product Code. 30136316
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136316 25 mg 25mg
30132989 10 mg 10mg
17954475 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136316 Supplier Toronto Research Chemicals Supplier No. TRCI49500025MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 482-44-0
Molecular Formula C16 H14 O4
Molecular Weight (g/mol) 270.28
Synonym 7H-Furo[3,2-g][1]benzopyran-7-one, 9-[(3-methyl-2-buten-1-yl)oxy]-, 7H-Furo[3,2-g][1]benzopyran-7-one, 9-[(3-methyl-2-butenyl)oxy]- (8CI,9CI), Imperatorin (6CI), 8-Isopent-2-enyloxy-6,7-furanocoumarin, 8-Isopentenyloxypsoralene, Ammidin, Marmelide, Marmelosin, NSC 402949
IUPAC Name 9-(3-methylbut-2-enoxy)furo[3,2-g]chromen-7-one
SMILES CC(=CCOc1c2OC(=O)C=Cc2cc3ccoc13)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H14O4/c1-10(2)5-7-19-16-14-12(6-8-18-14)9-11-3-4-13(17)20-15(11)16/h3-6,8-9H,7H2,1-2H3
Formula Weight 270.0892
Chemical Name or Material Imperatorin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.