missing translation for 'onlineSavingsMsg'
Learn More

Incandronate, TRC

Product Code. 30137036
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137036 50 mg 50mg
30133959 100 mg 100mg
30010757 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137036 Supplier Toronto Research Chemicals Supplier No. TRCI49970050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124351-85-5
Molecular Formula C8H19NO6P2
Molecular Weight (g/mol) 287.19
Synonym P,P'-[(Cycloheptylamino)methylene]bisphosphonic Acid, [(Cycloheptylamino)methylene]bisphosphonic Acid, Cimadronate, Cimadronic Acid, Icandronate, Incadronic Acid, [(Cycloheptylamino)methylene]bis(phosphonic acid), [(Cycloheptylamino)methylene]diphosphonic Acid
IUPAC Name [(cycloheptylamino)-phosphonomethyl]phosphonic acid
SMILES OP(=O)(O)C(NC1CCCCCC1)P(=O)(O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H19NO6P2/c10-16(11,12)8(17(13,14)15)9-7-5-3-1-2-4-6-7/h7-9H,1-6H2,(H2,10,11,12)(H2,13,14,15)
Formula Weight 287.0688
Chemical Name or Material Incandronate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.