Learn More
OHAUS™ Incubating Shaker
Product Code. 17268380 Shop All Ohaus Products
Change view
Click to view available options
Speed Range:
15 to 500 rpm
500 to 2500 rpm
600 to 2500 rpm
Load Bearing Capacity (Metric):
16 kg at 75 rpm, 2.3 kg at 500 rpm
22.7 kg at 125 rpm, 4.53 kg at 500 rpm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Load Bearing Capacity (Metric) Unit Size
17268380 15 to 500 rpm 22.7 kg at 125 rpm, 4.53 kg at 500 rpm Each
17258380 600 to 2500 rpm 16 kg at 75 rpm, 2.3 kg at 500 rpm Each
17278380 500 to 2500 rpm 22.7 kg at 125 rpm, 4.53 kg at 500 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17268380 Supplier OHAUS™ Supplier No. 30573775
  • ONLINE EXCLUSIVE PRICE
    10900.00€
    7965.67€ / Pack of 1
    Save 2934.33€
    26% Off
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Save Incubator Space with Benchtop Incubating and Incubating Refrigerating Shakers
  • The exclusive Opti-Flow Forced Ventilation System utilizes twin induction or a single large fan
  • Air is deflected around & not directly on samples, creating a circular flow of air ensuring uniformity
  • For repeatable results, the exclusive Accu-Drive shaking system ensures speed accuracy
  • The Triple Eccentric Drive provides consistent shaking motion throughout the entire speed range
  • The chambers for the ISHD23HDG and ISHD23CDG units are large enough to hold 2 x 6 L Erlenmeyer flasks with optional clamps and platform.
TRUSTED_SUSTAINABILITY
Specifications
Load Bearing Capacity (Metric) 22.7 kg at 125 rpm, 4.53 kg at 500 rpm
Speed Range 15 to 500 rpm
Voltage 230 V
Temperature Range (Metric) 15°C to 32°C
Wattage 800 W
Load Bearing Capacity (English) 50 lb. at 125 rpm, 10 lb. at 500 rpm
Certifications/Compliance TUV
Frequency 50/60 Hz
Material Aluminum, Cycoloy
Dimensions (L x W x H) 815 x 675 x 596 mm
Height (English) 23.5 in.
Length (English) 32.1 in.
Width (English) 26.6 in.
Shipping Weight (Metric) 103.4 kg
Weight (English) 224.07 lb.
Weight (Metric) 101.6 kg
Model ISHD23HDG
Display Independent LED displays for temperature, speed and time
For Use With (Application) Cell Cultures, Solubility Studies, Bacterial Suspensions, Extraction Procedures
Humidity Range 80%, Non-condensing
Timer Range 1 sec. to 160 hr.