missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) chloride, 98%, BLD Pharm™

Product Code. 31033233
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31033233 10 g 10g
31025468 5 g -
31022310 25 g -
31019003 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31033233 Supplier BLD Pharm Supplier No. BD13458510g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 10025-82-8
Quantity 10 g
Molecular Formula Cl3In
MDL Number MFCD00011058
SMILES [In+3].[Cl-].[Cl-].[Cl-]
Molecular Weight (g/mol) 221.18
Percent Purity 98%
Chemical Name or Material Indium(III) chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.