Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2 with Certificate
Product Code. 15535189 Shop All Fisherbrand Products
Change view
Click to view available options
Weight (Metric):
1 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
1 unit
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) unitSize
15535189 10 kg 1 unit
15535069 50 g 1 unit
15575069 1 kg 1 unit
15585059 1 g 1 unit
15585069 2 kg 1 unit
15595069 5 kg 1 unit
15545069 100 g 1 unit
15555069 200 g 1 unit
15565069 500 g 1 unit
15525069 20 g 1 unit
15515069 10 g 1 unit
15505069 5 g 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15535189
  • ONLINE EXCLUSIVE PRICE
    1300.00€
    1105.00€ / 1 units
    Save 195.00€
    15% Off
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
  • Class E2 stainless steel calibration weight
  • In compliance with OIML R111
  • Delivered with a calibration certificate guaranteeing traceability
Specifications
Classification E2
Material Stainless Steel
Weight (Metric) 10 kg
Mass 10 kg
Shape Cylindrical