missing translation for 'onlineSavingsMsg'
Learn More

Indoprofen, TRC

Product Code. 30011049
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011049 25 mg 25mg
17949829 10 mg 10mg
17929719 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011049 Supplier Toronto Research Chemicals Supplier No. TRCI64500025MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31842-01-0
Molecular Formula C17 H15 N O3
Molecular Weight (g/mol) 281.31
Synonym Benzeneacetic acid, 4-(1,3-dihydro-1-oxo-2H-isoindol-2-yl)-α-methyl-, (±)-Indoprofen, 1-Oxo-2-[p-[(α-methyl)carboxymethyl]phenyl]isoindoline, 2-[4-(1-Oxo-2-isoindolinyl)phenyl]propanoic acid, Bor-ind, Flosin, Flosint, IPP, Indoprofen, Isindone, K 4277, Praxis, Racemic Indoprofen, Reumofene, dl-α-[4-(1-Oxo-2-isoindolinyl)phenyl]propionic acid, p-(1-Oxo-2-isoindolinyl)hydratropic acid
IUPAC Name 2-[4-(3-oxo-1H-isoindol-2-yl)phenyl]propanoic acid
SMILES CC(C(=O)O)c1ccc(cc1)N2Cc3ccccc3C2=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H15NO3/c1-11(17(20)21)12-6-8-14(9-7-12)18-10-13-4-2-3-5-15(13)16(18)19/h2-9,11H,10H2,1H3,(H,20,21)
Formula Weight 281.1052
Chemical Name or Material Indoprofen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.