missing translation for 'onlineSavingsMsg'
Learn More

Indoramin EP Impurity A, Mikromol™

Product Code. 30049074
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049074 100 mg 100mg
30052527 25 mg 25mg
30459651 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30049074 Supplier Mikromol Supplier No. MM0802.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Indoramin EP Impurity A
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(2-Bromoethyl)indole
CAS 3389-21-7
Analyte or Component Names 3-(2-Bromoethyl)indole
Shipping Condition Room Temperature
Molecular Formula C10 H10 Br N
Certifications/Compliance ISO 9001
Synonym 3-(2-Bromoethyl)indole, Indoramin Hydrochloride Imp. A (BP), Indoramin Hydrochloride Impurity A, Indoramin Impurity A
InChI Formula InChI=1S/C10H10BrN/c11-6-5-8-7-12-10-4-2-1-3-9(8)10/h1-4,7,12H,5-6H2
SMILES BrCCc1c[nH]c2ccccc12
IUPAC Name 3-(2-bromoethyl)-1H-indole
Molecular Weight (g/mol) 224.1
Quantity 100 mg
Formula Weight 222.9997 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.