missing translation for 'onlineSavingsMsg'
Learn More

Indoxacarb, TRC

Product Code. 30011610
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011610 25 mg 25mg
30004379 250 mg 250mg
30011231 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011610 Supplier Toronto Research Chemicals Supplier No. TRCI65401025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 173584-44-6
Molecular Formula C22 H17 Cl F3 N3 O7
Molecular Weight (g/mol) 527.83
Synonym Indeno[1,2-e][1,3,4]oxadiazine-4a(3H)-carboxylic acid, 7-chloro-2,5-dihydro-2-[[(methoxycarbonyl)[4-(trifluoromethoxy)phenyl]amino]carbonyl]-, methyl ester, (4aS)-, Indeno[1,2-e][1,3,4]oxadiazine-4a(3H)-carboxylic acid, 7-chloro-2,5-dihydro-2-[[(methoxycarbonyl)[4-(trifluoromethoxy)phenyl]amino]carbonyl]-, methyl ester, (S)-, (S)-Indoxacarb, Activyl, Advion, Advion fire ant bait, Avatar, Avaunt, Avaunt 30WDG, Avaunt 30WG, DPX-KN 128, DPX-MP 062-381, DPX-MP 062-382, DPX-MP 062-385, DPX-MP 062-390, DPX-MP 062-391, DPX-MP 062-400, Explicit, Indoxacarb, Indoxacarb MP, KN 128, Parkway Mole Cricket Bait, Provaunt, S-(+)-Indoxacarb, Steward, Steward 30WG, Steward EC, Steward Gold
IUPAC Name methyl (4aS)-7-chloro-2-[methoxycarbonyl-[4-(trifluoromethoxy)phenyl]carbamoyl]-3,5-dihydroindeno[1,2-e][1,3,4]oxadiazine-4a-carboxylate
SMILES COC(=O)N(C(=O)N1CO[C@]2(Cc3cc(Cl)ccc3C2=N1)C(=O)OC)c4ccc(OC(F)(F)F)cc4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H17ClF3N3O7/c1-33-18(30)21-10-12-9-13(23)3-8-16(12)17(21)27-28(11-35-21)19(31)29(20(32)34-2)14-4-6-15(7-5-14)36-22(24,25)26/h3-9H,10-11H2,1-2H3/t21-/m0/s1
Formula Weight 527.0707
Percent Purity >95%
Chemical Name or Material (S)-Indoxacarb
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.