Learn More
Terracycle™ Industrial Safety Equipment Zero Waste Box™
Product Code. 17865735
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17865735 Small Each
17855925 Medium Each
17805715 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17865735 Supplier Terracycle™ Supplier No. CS199SBE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Recycle Industrial Safety Equipment using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
Use this Zero Waste Box to recycle:
  • Industrial safety helmets
  • Safety bump caps
  • Hi-vis jackets
  • Heavy-duty gloves
  • Ear defenders Protective eyewear (goggles)
Do not send in:
  • Fabric work gloves with an anti-slip layer
  • Suede and leather gloves
  • Disposable face masks
  • Safety footwear
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
  • Batteries, pressurized canisters, broken glass, and medical waste
Specifications
Capacity (Metric) 7 kg
Capacity (English) 15 lb.
Color Beige, Green, Red, White
Dimensions (L x W x H) 25 x 25 x 46 cm
Disposable Single-use
Latex Free Yes
Size Small
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box