missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 M2 Antibody - (A/Puerto Rico/8/1934), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14997
This item is not returnable.
View return policy
Description
Influenza A H1N1 M2 Polyclonal antibody specifically detects Influenza A H1N1 M2 in Virus samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
| Influenza A H1N1 M2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Influenza A virus. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| H1N1 | |
| Recombinant protein corresponding to Influenza A virus M2 (matrix protein) (A/Puerto Rico/8/1934(H1N1). The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction