missing translation for 'onlineSavingsMsg'
Learn More

Influenza B Hemagglutinin Antibody (10A6E9C7) - (B/Phuket/3073/2013), Novus Biologicals™

Mouse Monoclonal Antibody

265.00€ - 2380.00€

Specifications

Antigen Influenza B Hemagglutinin
Clone 10A6E9C7
Dilution ELISA 1:1000-1:2000, Sandwich ELISA Capture 1:250-1:2000
Applications ELISA, Sandwich ELISA Capture
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18695527
View Documents
Novus Biologicals
NBP3-06570-100ul
100 μg
459.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18662769
View Documents
Novus Biologicals
NBP3-06570-1ml
1 mL
2380.00€
1mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18620897
View Documents
Novus Biologicals
NBP3-06570-50ul
50 μg
265.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Influenza B Hemagglutinin Monoclonal antibody specifically detects Influenza B Hemagglutinin in Virus samples. It is validated for ELISA, Sandwich ELISA Capture
Specifications

Specifications

Influenza B Hemagglutinin
ELISA 1:1000-1:2000, Sandwich ELISA Capture 1:250-1:2000
Monoclonal
Purified
RUO
0.2 um filtered solution in PBS
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza B Hemagglutinin (Accession#: ACA33493.1; Met 1-Ala 555).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
10A6E9C7
ELISA, Sandwich ELISA Capture
Unconjugated
Mouse
Virus
B1AG46
IgG2b
Protein A purified
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.