Learn More
TFA Dostmann Infrared Thermometer SLIM FLASH
Product Code. 17991312
Change view
Click to view available options
Calibrated:
Yes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Calibrated Unit Size
17991312 Yes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17991312 Supplier TFA Dostmann Supplier No. 31.1128
  • ONLINE EXCLUSIVE PRICE
    24.05€
    18.81€ / Pack of 1
    Save 5.24€
    21% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The SLIM FLASH infrared thermometer can measure the temperature of surfaces without touching them. Simply aim at the object to be measured, press the button and the result is obtained in seconds. The handy device is reminiscent of a USB stick and is as well HACCP-compliant.
With the SLIM FLASH infrared thermometer from TFA, you can quickly and easily measure the temperature of surfaces without touching them. Simply aim at the object to be measured, press the button and the result is obtained in seconds. The handy device is reminiscent of a USB stick and fits in any trouser pocket or on a key ring. Infrared thermometers can be used for a wide range of applications. You can use them to check air conditioning and heating systems, look for cold bridges and faulty relays or check the brake system in your car. In the home, you can check the temperature of baby food, drinks, bath water, frying fat, jam or chocolate mass. The device is HACCP-compliant and the hygienic, contact-free temperature measurement is particularly suitable for professional use in the food sector. Non-contact measurement - made possible by proven infrared technology. The infrared radiation emitted by any object with a temperature above absolute zero is converted into an electrical signal, evaluated by a processor and then shown on the display.
  • Contact-free measuring of surface temperature
  • Small, easy to use
  • Compliant with HACCP, ideal for food
  • A versatile tool for the household, craft and hobby
  • Response time 1 sec.
  • precision +- 2% or 2°C
  • Registered TFA design
Specifications
Calibrated Yes
Display LCD