Learn More
METTLER TOLEDO™ InLab™ Expert pH Electrodes
Product Code. 11394203
Change view
Click to view available options
Connector Types:
BNC, RCA
BNC, RCA IP67
DIN 19262, 4 mm Banana Plug
Multi-Pin
S7
Includes:
Electrode only
ISM Temperature Probe
Pt1000 Temperature Probe
Temperature Probe
Temperature Probe (NTC30)
Length (Metric) Cable:
1.2 m
1.8 m
10 m
2 m
No Cable
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Includes Length (Metric) Cable Unit Size
11394203 BNC, RCA ISM Temperature Probe 1.2 m Each
10454353 Multi-Pin Temperature Probe (NTC30) No Cable Each
10009552 S7 Electrode only No Cable Each
10610904 DIN 19262, 4 mm Banana Plug Pt1000 Temperature Probe 1.2 m Each
10040073 BNC, RCA ISM Temperature Probe 2 m Each
10454543 Multi-Pin Pt1000 Temperature Probe No Cable Each
10660324 BNC, RCA Temperature Probe 1.2 m Each
10447144 BNC, RCA IP67 Temperature Probe 2 m Each
10431435 BNC, RCA IP67 ISM Temperature Probe 1.8 m Each
10594925 BNC, RCA IP67 ISM Temperature Probe 10 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11394203 Supplier METTLER TOLEDO™ Supplier No. 30014096
  • ONLINE EXCLUSIVE PRICE
    472.75€
    419.01€ / Pack of 1
    Save 53.74€
    11% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.
Ideal for critical samples such as suspensions or samples of unknown composition. Mettler Toledo Expert Series pH and pH/ATC Combination Electrodes with open junction puts the reference electrode in direct contact with the measured medium, eliminating the possibility of contamination or blockage.
  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Features a PEEK plastic shaft for harsh environments and requires no maintenance
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
  • General Lab
  • Beverages
  • Waste water
  • Suspended solids
  • Environmental
  • TRIS compatible
Accessories & Care
  • Open Junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC, RCA
Junction Open
Material PEEK
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology
Filling Solution Type Xerolyt Polymer
For Use With (Application) Emulsions, suspensions, effluent water and solutions containing Proteins and Sulphides
For Use With (Equipment) Fixed Cable to sensor
Includes ISM Temperature Probe
Internal Reference ARGENTHAL
Membrane U Glass
Model InLab Expert Pro-ISM
Probe NTC 30 kilohm
Response Time <20 sec.
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.7 in.
Length (English) Cable 47.2 in.
Length (Metric) 120 mm
Length (Metric) Cable 1.2 m
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm