Learn More
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection
Product Code. 15529661 Shop All Mettler Toledo Products
Click to view available options
Connector Types:
S7-BNC
S7-BNC, IP67 waterproof
S7-DIN 19262
S7-Lemo 00
S7-None
Length (Metric) Cable:
1.2 m
10 m
3.0 m
5.0 m
Unit Size:
Each
Product Code. 15529661
Brand: Mettler Toledo™ 30281921
Only null left
Add to basket
Add to basket
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter.
  • Detachable
  • Resuable
TRUSTED_SUSTAINABILITY
Specifications
Connector Types S7-DIN 19262
For Use With (Equipment) Mettler Toledo InLab Sensors
For Use With (Application) To connect Mettler Toledo InLab sensors to any meter
Length (English) Cable 16.4 ft.
Length (Metric) Cable 5.0 m