missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ InnovaCoat (R) Gold - 10nm Midi Maleimide Antibody Labeling Kit

Product Code. 18093326 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18093326 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18093326 Supplier Novus Biologicals™ Supplier No. 2720015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody labeling kit

This conjugation kit allows thiolated molecules (e.g. antibodies, Fab’ fragments, or other sulfhydryl-containing biomolecules) to be covalently attached to ultra-stable InnovaCoat™ GOLD nanoparticles at very high OD quickly and easily.

Kit Contents:

  • 1 vial (1 reaction) of InnovaCoat™ GOLD Maleimide
  • 1 vial of InnovaCoat™ GOLD Maleimide Reaction Buffer
  • 1 vial of InnovaCoat™ GOLD Maleimide Quencher
  • 1 vial or bottle of InnovaCoat™ GOLD Maleimide Diluent
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 400 μL
Target InnovaCoat (R) Gold - 10nm Midi Maleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.