Learn More
WTW™ inoLab™ Cond 7310™ Precise and Convenient Conductivity Meter
GREENER_CHOICE
Product Code. 12078846
Change view
Click to view available options
Type:
Conductivity Meter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
12078846 Conductivity Meter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 12078846 Supplier WTW™ Supplier No. 1CA300
Only null left
Add to basket
Add to basket
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Precise and convenient conductivity meter product type that includes meter with universal power supply, stand and operation manual, software and USB cable.
  • Precise and convenient conductivity meter product type
  • Includes meter with universal power supply, stand and operation manual, software and USB cable
TRUSTED_SUSTAINABILITY
Specifications
Includes Meter with universal power supply, stand and operation manual, software and USB cable
Accuracy (Conductivity) ±0.5% of value
Outputs 9V, 1.1A
Cell Constant Fixed 0.01cm–1, calibration 0.450 to 0.500cm–1, 0.800 to 0.880cm–1, adjustable 0.09 to 0.110cm–1, 0.250 to 25cm–1
Calibration Points 1
Range (Conductivity) 0 μS/cm to 1000 mS/cm
Temperature Compensation nLF, linear 0.000 to 10.000%, can be switched-off
Range (Salinity) 0 to 70 ppt
Range (TDS) 0 to 1999 mg/L
Inputs 100 to 240V, 50 to 60Hz, 270mA
Certifications/Compliance cETLus, CE
Weight (Metric) 0.8 kg
Model inoLab Cond 7310
Power Supply 4 x 1.5V Alkali Manganese batteries, Type AA, 4 x 1.2V NiMH rechargeable batteries, type AA
Temperature (Metric) Operating -5°C to +105°C
Type Conductivity Meter
Electrical Requirements 100 to 240V, 50/60Hz, 270mA input, 9V output
Voltage 100/240 V
Frequency 50/60Hz