missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inosine-3',5'-cyclic Monophosphate Sodium Salt, TRC
Specifications
Specifications
| CAS | 41092-64-2 |
| Molecular Formula | C10 H10 N4 O7 P . Na |
| Quantity | 10 mg |
| Synonym | Inosine Cyclic 3',5'-(Hydrogen Phosphate) Sodium Salt |
| InChI Formula | InChI=1S/C10H11N4O7P.Na/c15-6-7-4(1-19-22(17,18)21-7)20-10(6)14-3-13-5-8(14)11-2-12-9(5)16;/h2-4,6-7,10,15H,1H2,(H,17,18)(H,11,12,16);/q;+1/p-1/t4-,6-,7-,10-;/m1./s1 |
| SMILES | [Na+].O[C@@H]1[C@@H]2OP(=O)([O-])OC[C@H]2O[C@H]1n3cnc4C(=O)NC=Nc34 |
| IUPAC Name | sodium;9-[(4aR,6R,7R,7aS)-7-hydroxy-2-oxido-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl]-1H-purin-6-one |
| Molecular Weight (g/mol) | 352.17 |
| Formula Weight | 352.0185 |
| Chemical Name or Material | Inosine-3',5'-cyclic Monophosphate Sodium Salt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?