missing translation for 'onlineSavingsMsg'
Learn More

Inosine 5’ Triphosphate Trisodium Salt, TRC

Product Code. 30179892
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179892 250 mg 250mg
30180171 100 mg 100mg
30179802 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179892 Supplier Toronto Research Chemicals Supplier No. TRCI665000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35908-31-7
Molecular Formula C10 H12 N4 O14 P3 . 3 Na
Molecular Weight (g/mol) 574.111
Synonym Inosine 5'-(Tetrahydrogen Triphosphate) Trisodium Salt, NSC 20266, Trisodium Inosine-5'-triphosphate;
IUPAC Name trisodium;[[[(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-1H-purin-9-yl)oxolan-2-yl]methoxy-oxidophosphoryl]oxy-oxidophosphoryl] hydrogen phosphate
SMILES [Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])OP(=O)([O-])OP(=O)(O)[O-])n2cnc3C(=O)NC=Nc23

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H15N4O14P3.3Na/c15-6-4(1-25-30(21,22)28-31(23,24)27-29(18,19)20)26-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;;/h2-4,6-7,10,15-16H,1H2,(H,21,22)(H,23,24)(H,11,12,17)(H2,18,19,20);;;/q;3*+1/p-3/t4-,6-,7-,10-;;;/m1.../s1
Formula Weight 573.926
Chemical Name or Material Inosine 5' Triphosphate Trisodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.