missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSM1 Antibody (CL11630) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-15120UV
This item is not returnable.
View return policy
Description
INSM1 Monoclonal antibody specifically detects INSM1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| INSM1 | |
| Monoclonal | |
| DyLight 350 | |
| IA1IA-1, insulinoma-associated 1, insulinoma-associated protein 1, Zinc finger protein IA-1 | |
| This antibody was developed using a synthetic peptide derived from Q01101, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 3642 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL11630 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction