missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INSR beta Recombinant Rabbit Monoclonal Antibody (HL2737)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556762
This item is not returnable.
View return policy
Description
INSR beta Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Biological actions of insulin and IGF-1 are mediated by their respective cell surface receptor tyrosine kinases that regulate multiple signaling pathways through activation of a series of phosphorylation cascades. The insulin receptor. Insulin/IGF-1 binding to the extracellular domain leads to autophosphorylation of downstream target proteins. These two receptors differ in sequence in regions that confer specificity for the designated ligand as well as in certain intracellular signaling domains, resulting in significant differences in the functional consequences of activation of each receptor. Defects in IR are the cause of various insulin resistance syndromes and IGF-1R defects may cause some forms of growth retardation. Both these signaling cascades are also important for the development of cancer.
Specifications
| INSR beta | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P06213, P15208 | |
| INSR | |
| Recombinant fragment of human Insulin Receptor beta. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2737 | |
| Unconjugated | |
| INSR | |
| 4932439J01Rik; CD220; CD220 beta; CD221; D630014A15Rik; HHF5; IGF-I receptor; Insr; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR beta; IR-A; IR-B | |
| Rabbit | |
| Protein A | |
| RUO | |
| 16337, 3643 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction