Learn More
Binder Installation ICH Light Module
Product Code. 30503400 Shop All BINDER Products
Change view
Click to view available options
Duration:
1 Day
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Duration unitSize
30503400 1 Day 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30503400 Supplier Binder Supplier No. DL100140
Installation of the BINDER ICH-Q1B light module.
  • Mount the supply box, install and connect the light cassettes, (LQC: and light sensors,) seal the access port, (LQC: update the controller dataset), and check function
Specifications
For Use With (Equipment) BINDER KB PRO/KBF/KBF PRO and ICH Light Module
Duration 1 Day
Product Type Installation Remote Support