Learn More
Speirs Robertson™ Instrument Isolation Platform
Product Code. 11969434
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
30 x 45 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11969434 30 x 45 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11969434 Supplier Speirs Robertson™ Supplier No. AMDSR 90x75SS
Estimated Shipment: 11-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Improve performance of sensitive instruments by shielding them from vibrations. Speirs Robertson™ Instrument Isolation Platform is simple to install, easy to use, and maintenance free.
  • Sorbothane isolators
  • Improve instrument performance by removing vibrations
  • Suitable for instruments weighing up to 25kg
  • Combination of isolators and sound-deadening, layered construction
  • Seamless pharma-grade (Ra finish 0.3) stainless steel worktop
  • Chemically resistant and easy to clean
Specifications
Dimensions (L x W) 30 x 45 mm